missing translation for 'onlineSavingsMsg'
Learn More

Xanthan Gum, TCI America™

Catalog No. p-7137964
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X004825G 25 g
X0048500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X004825G Supplier TCI America Supplier No. X004825G
Only null left

Chemical Identifiers

CAS 11138-66-2
Molecular Formula C8H14Cl2N2O2
Molecular Weight (g/mol) 241.112
MDL Number MFCD00131256
InChI Key VXYWXJXCQSDNHX-UHFFFAOYSA-N
Synonym 2-2,4-diaminophenoxy ethanol dihydrochloride, 2,4-diaminophenoxyethanol dihydrochloride, 2,4-diaminophenoxyethanol hcl, unii-z5y843j6kq, ethanol, 2-2,4-diaminophenoxy-, dihydrochloride, 2,4-diaminophenoxyethanol 2hcl, 2-2,4-diaminophenoxy ethanol hydrochloride, ethanol, 2-2,4-diaminophenoxy-, hydrochloride 1:2, 4-2-hydroxyethoxy-1,3-phenylenediamine dihydrochloride, 2,4-diaminophenoxyethanol hydrochloride
PubChem CID 47932
IUPAC Name 2-(2,4-diaminophenoxy)ethanol;dihydrochloride
SMILES C1=CC(=C(C=C1N)N)OCCO.Cl.Cl

Specifications

Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Xanthan Gum