missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xanthan Gum, NF, 91-108%, Spectrum™ Chemical
XA105, 11138-66-2
$708.30
Chemical Identifiers
| CAS | 11138-66-2 |
|---|---|
| Molecular Formula | C8H14Cl2N2O2 |
| Molecular Weight (g/mol) | 241.11 |
| InChI Key | VXYWXJXCQSDNHX-UHFFFAOYSA-N |
| IUPAC Name | 2-(2,4-diaminophenoxy)ethan-1-ol dihydrochloride |
Description
Spectrum™ Chemical Xanthan Gum, NF is a natural gum polysaccharide used as a thickener or rheology modifier and emulsion stabilizer. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 11138-66-2 | |
| 241.11 | |
| 2-(2,4-diaminophenoxy)ethan-1-ol dihydrochloride |
| C8H14Cl2N2O2 | |
| VXYWXJXCQSDNHX-UHFFFAOYSA-N |
Specifications
| 11138-66-2 | |
| VXYWXJXCQSDNHX-UHFFFAOYSA-N | |
| 241.11 | |
| NF |
| C8H14Cl2N2O2 | |
| 2-(2,4-diaminophenoxy)ethan-1-ol dihydrochloride | |
| 91 to 108% |