missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xanthan Gum, FCC, 4.2-5.4%, Spectrum™ Chemical
X1001, 11138-66-2
Supplier: Spectrum Chemical Mfg Cor X1001500GM
| Quantity | 500 g |
|---|---|
| Packaging | Poly Bottle |
Description
Spectrum™ Chemical Xanthan Gum, FCC is a polysaccharide most often used as a food thickening agent and stabilizer. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 11138-66-2 | |
| VXYWXJXCQSDNHX-UHFFFAOYSA-N | |
| Cl.Cl.NC1=CC(N)=C(OCCO)C=C1 |
| 241.11 | |
| 2-(2,4-diaminophenoxy)ethan-1-ol dihydrochloride |
Specifications
| 11138-66-2 | |
| 500 g | |
| Cl.Cl.NC1=CC(N)=C(OCCO)C=C1 | |
| 241.11 | |
| 4.2 to 5.4% |
| Poly Bottle | |
| VXYWXJXCQSDNHX-UHFFFAOYSA-N | |
| 2-(2,4-diaminophenoxy)ethan-1-ol dihydrochloride | |
| To pass test | |
| FCC |