missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ X-Y Axis Dry Erase Mat
Double sided dry-erase mat includes a 1 cm x-y axis grid on one side for graphing activities.
Supplier: United Scientific™ UNCG9X11PK30
Alert:
Prop 65
Description
- Double sided dry-erase mat
- Includes a 1 cm x-y axis grid on one side for graphing activities.
- The other side is blank but coated so it can be used with dry-erase markers.
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Graphing Sheets | |
30 Pk. | |
9 x 11 in. |