missing translation for 'onlineSavingsMsg'
Learn More

Poltex Wypall Dispenser X60

Dispenser holds and dispenses Wypall x60, #34790 wipes. Durable PETG construction.

Supplier:  Poltex WYPL34790T

 View more versions of this product

Catalog No. 22022529


Only null left

Description

Description

  • Holds and dispenses Wypall x60, #34790 wipes
  • Durable PETG construction
  • Available as magnet mount (powerful magnets included), wall mount (screws/anchors included), and tape mount (VHB tape included)
Specifications

Chemical Identifiers

Clear
248.92 mm
Wipes, Labwipes
1
Non-sterile
127 mm
248.92 mm
PETG
Side Loading

Specifications

Wipe Dispenser
127 mm
248.92 mm
PETG
1
Non-sterile
Clear
248.92 mm
Wipes, Labwipes
VHB Tape
Side Loading
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions