missing translation for 'onlineSavingsMsg'
Learn More

WTAP Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501560386
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501560386 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501560386 Supplier Bethyl Laboratories Inc Supplier No. A301436A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with X. laevis and X. tropicalis

The Wilms tumor suppressor gene WT1 appears to play a role in both transcriptional and posttranscriptional regulation of certain cellular genes. This gene encodes a WT1-associating protein, which is a ubiquitously expressed nuclear protein. Like WT1 protein, this protein is localized throughout the nucleoplasm as well as in speckles and partially colocalizes with splicing factors. Alternative splicing of this gene results in three transcript variants, two of which encode the same isoform.

Specifications

Antigen WTAP
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene WTAP
Gene Accession No. Q15007
Gene Alias PNAS-132, WT1-associated protein, Wilms′ tumour 1-associating protein, female-lethal(2)D homolog, hFL(2)D, pre-mRNA-splicing regulator WTAP, putative pre-mRNA splicing regulator female-lethal(2D), wilms tumor 1-associating protein
Gene Symbols WTAP
Host Species Rabbit
Immunogen Between 346 and 396
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9589
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less