missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Write On Right-To-Know Laboratory Labels, Vinyl

Catalog No. 19110332
Change view
Click to view available options
Color:
Black, Blue, Red, Yellow, White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19110332 Black, Blue, Red, Yellow, White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19110332 Supplier Brady™ Supplier No. 121161

RTK labels use color coding system in conjunction with written text to identify chemical, physical and health hazards. Brady Write On Right-To-Know Laboratory Labels allow user to customize message to suit needs.

  • To identify the chemical and its physical and health hazards
  • Allows to customize message to suit needs
  • Quantity: 500 per roll

Specifications

Description HMIG Color Bar
Color Black, Blue, Red, Yellow, White
Finish Gloss
Material Vinyl (B-7569)
Product Type Write On Right-To-Know Laboratory Label
Display Text Legend: HEALTH HAZARD FLAMMABILITY HAZARD INSTABILITY HAZARD PERSONAL PROTECTION
Quantity 500/Roll
Shape Rectangle
Size (English) 1.25 x 2 in. (W x H)
Size (Metric) 31.7 x 50.8 mm (W x H)