missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Write-On Magnetic Labels

Catalog No. 19112916
Change view
Click to view available options
Color:
Blue
Green
Orange
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19112916 Blue
19112917 Green
19112918 Orange
19112921 Yellow
19112919 Red
19112920 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19112916 Supplier Brady™ Supplier No. 56957

Label and update with incredible flexibility. Brady™ Write-On Magnetic Labels can be used for flexible short-term labeling with the included water-soluble pen, or can be used for permanent labeling with a permanent marking pen.

  • Re-useable labels with water-soluble pen (included) that wipes off with wet cloth
  • Special label surface accepts water-soluble pen
  • Magnetic material available in six colors
  • Sold on 1 in. x 50 ft. rolls

Specifications

Color Blue
Format Roll
Material Magnetic
Product Type Write-on Magnetic Labels
Shape Rectangle
Size (English) 50 ft. x 1 in.
Size (Metric) 15.2 m x 2.54 cm