missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wright's Stain, Rapid Formula, for Staining and General Differentiation of Blood Corpuscles, Ricca Chemical
$180.28 - $180.28
Chemical Identifiers
| CAS | 67-56-1 |
|---|---|
| Molecular Formula | HNa2O4P |
| Molecular Weight (g/mol) | Mixture |
| MDL Number | MFCD00003496 |
| InChI Key | BNIILDVGGAEEIG-UHFFFAOYSA-L |
| PubChem CID | 24203 |
| ChEBI | CHEBI:34683 |
| IUPAC Name | disodium hydrogen phosphate |
| SMILES | [Na+].[Na+].OP([O-])([O-])=O |
Chemical Identifiers
| 67-56-1 | |
| Mixture | |
| BNIILDVGGAEEIG-UHFFFAOYSA-L | |
| CHEBI:34683 | |
| [Na+].[Na+].OP([O-])([O-])=O |
| HNa2O4P | |
| MFCD00003496 | |
| 24203 | |
| disodium hydrogen phosphate |
Specifications
| 67-56-1,56-81-5,68988-92-1,7778-77-0,51811-82-6,7558-79-4 | |
| MFCD00003496 | |
| BNIILDVGGAEEIG-UHFFFAOYSA-L | |
| disodium hydrogen phosphate | |
| 24203 | |
| Laboratory | |
| Amber Glass Bottle | |
| 6.30-6.50 | |
| Solution |
| HNa2O4P | |
| Solubility in Water: Infinite | |
| [Na+].[Na+].OP([O-])([O-])=O | |
| Mixture | |
| CHEBI:34683 | |
| Characteristic alcohol | |
| Dark Purple | |
| 500 mL | |
| Wright's Stain |
Safety and Handling
Recommended Storage : Flammable