missing translation for 'onlineSavingsMsg'
Learn More

Wright's Stain, Rapid Formula, for Staining and General Differentiation of Blood Corpuscles, Ricca Chemical

Supplier:  Ricca Chemical Company 935016

 View more versions of this product

Catalog No. 935016


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Wright's Stain
HNa2O4P
Solubility in Water: Infinite
[Na+].[Na+].OP([O-])([O-])=O
Mixture
CHEBI:34683
Characteristic alcohol
Dark Purple
500 mL
67-56-1,56-81-5,68988-92-1,7778-77-0,51811-82-6,7558-79-4
MFCD00003496
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium hydrogen phosphate
24203
Laboratory
Amber Glass Bottle
6.30-6.50
Solution

Chemical Identifiers

67-56-1
Mixture
BNIILDVGGAEEIG-UHFFFAOYSA-L
CHEBI:34683
[Na+].[Na+].OP([O-])([O-])=O
HNa2O4P
MFCD00003496
24203
disodium hydrogen phosphate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Flammable

SDS Documents

Documents

Promotions

Promotions