missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Wraptor Nylon Cloth Labels
Description
- Made from premium grade B-946 vinyl
- Sized to fit standard Blank Write-On Right-To-Know Labels
- Correspond to ANSI proposed Z.535 Standard
- Card-mounted for easy removal
Specifications
Specifications
| Certifications/Compliance | UL Recognized, CSA Accepted, Halogen Free (DIN VDE 0472/part 815) |
| Description | Suggested Ribbon Series: R4300 |
| Adhesive Type | Acrylic, Permanent, Cold-temperature |
| Color | White |
| Finish | Matte |
| Format | Roll |
| Material | Nylon Cloth |
| Print Technology | Thermal Transfer |
| Product Type | Wraptor Label |
| For Use With (Application) | Arc Flash Labels; Arrow Tape; Pipe and Valve Marking; Safety and Compliance ID; Visual Workplace Labeling; Warehouse Marking |
| Show More |