missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WNT3 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518516
Description
This antibody is predicted to react with canine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
WNT3, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.Chemical Identifiers
| WNT3 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| WNT3 | |
| INT4; Int-4; MGC131950; MGC138321; MGC138323; Proto-oncogene Int-4; proto-oncogene Int-4 homolog; proto-oncogene protein Wnt-3; proto-oncogene Wnt-3; TETAMS; wingless-related MMTV integration site 3; Wingless-type MMTV integration site 3, homolog; wingless-type MMTV integration site family member 3; wingless-type MMTV integration site family, member 3; Wnt family member 3; Wnt3; Wnt-3; WNT-3 proto-oncogene protein | |
| WNT3 | |
| Primary | |
| Ammonium sulfate precipitation |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Goat | |
| RUO | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P56703 | |
| 7473 | |
| Synthetic peptide sequence (CGRGHNTRTEKRKEK) corresponding to the internal amino acids of WNT3 (aa 315-330). | |
| Antibody |
Specifications
| WNT3 | |
| Polyclonal | |
| Unconjugated | |
| WNT3 | |
| INT4; Int-4; MGC131950; MGC138321; MGC138323; Proto-oncogene Int-4; proto-oncogene Int-4 homolog; proto-oncogene protein Wnt-3; proto-oncogene Wnt-3; TETAMS; wingless-related MMTV integration site 3; Wingless-type MMTV integration site 3, homolog; wingless-type MMTV integration site family member 3; wingless-type MMTV integration site family, member 3; Wnt family member 3; Wnt3; Wnt-3; WNT-3 proto-oncogene protein | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 7473 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P56703 | |
| WNT3 | |
| Synthetic peptide sequence (CGRGHNTRTEKRKEK) corresponding to the internal amino acids of WNT3 (aa 315-330). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |