missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WNT11 Monoclonal Antibody (D6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA566061
Description
WNT11 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.Specifications
| WNT11 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O96014 | |
| WNT11 | |
| Recombinant Wingless Type MMTV Integration Site Family, Member 11 (WNT11) (Ser204-Glu349). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| D6 | |
| Unconjugated | |
| WNT11 | |
| HWNT11; Protein Wnt-11; wingless-related MMTV integration site 11; wingless-type MMTV integration site family, member 11; Wnt family member 11; Wnt11; Wnt-11 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 7481 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |