missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WIZ Polyclonal Antibody

Description
A suggested positive control is rat lung tissue lysate. PA5-21082 can be used with blocking peptide PEP-1196.
WIZ, also known as ZNF803, was initially identified in a mouse cerebellum cDNA library screen and its message was found to be expressed in the granule cell layers of the cerebellum as well as in the dentate gyrus and olfactory bulb. Later analysis indicates however that WIZ is ubiquitously expressed. WIZ is a nuclear protein that co-localizes with G9a, a histone methyltransferase responsible for mono- and dimethylation of H3K9 at euchromatic regions. WIZ can also associate with CtBP family proteins, known to be transcriptional co-repressors, and has been suggested to link G9a/GLP complexes to the CtBP co-repressor machinery, possibly regulating complex stability and gene silencing.
Specifications
Specifications
| Antigen | WIZ |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | WIZ |
| Gene Accession No. | O88286, O95785 |
| Gene Alias | Protein Wiz; widely interspaced zinc finger motifs; widely-interspaced zinc finger motifs; widely-interspaced zinc finger-containing protein; Wiz; WIZ zinc finger; zinc finger protein 803; ZNF803 |
| Gene Symbols | WIZ |
| Show More |