missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Withaferin A, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 5119-48-2 |
| Flash Point | 226°C |
| Physical Form | Neat |
| Linear Formula | C28H38O6 |
| MDL Number | MFCD10687098 |
| Synonym | 4 beta,5 beta,6 beta,22 R)-5,6-Epoxy-4,22,27-trihydroxy-1-oxo-ergosta-2,24-dien-26-oic acid delta-lactone; Withaferine |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 470.60 |
| Percent Purity | ≥95% (HPLC) |
| Show More |