missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Thin Wall Quartz EPR Sample Tube
Manufactured from naturally occurring quartz with the highest purity to minimize the background signal. Bel-Art SP Scienceware™ Thin Wall Quartz EPR Sample Tube may contain Fe2O3 which is paramagnetic (1200 ppm) and has certain impact on shimming quality depending on the magnetic field strength.
$56.33 - $56.33
Specifications
| Length (Metric) | 250 mm |
|---|---|
| Material | Quartz |
| Product Type | Sample Tube |
| Thickness (Metric) Wall | 0.5 mm |
| Catalog Number | Mfr. No. | Diameter (Metric) Outer | Diameter (Metric) Inner | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (Metric) Outer | Diameter (Metric) Inner | Price | Quantity | ||||
|
16-800-114
|
Bel-Art™
660000014 |
4 mm | 3 mm |
Each for $56.33
|
|
||||
Description
- Tight ID and OD tolerances ensure 100% compatibility to spectrometers
- Individually wrapped to prevent contamination
- Recommended for experiments with small organic molecule (Molecular Weight ∼500) at ambient temperature only
Specifications
| 250 mm | |
| Quartz | |
| Sample Tube | |
| 0.5 mm |