missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wijs solution, Volumetric, 0.1 M ICl, Honeywell Fluka™
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
| Quantity | 1 L |
| Linear Formula | ICl |
| MDL Number | MFCD00036152 |
| UN Number | UN2920 |
| Beilstein | 3587194 |
| Flash Point | 40°C (104°F) |
| Synonym | iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french |
| Formula Weight | 162.36g/mol |
| Chemical Name or Material | Wijs solution |
Safety and Handling
| Hazard Statement | P280-P305 + P351 + P338-P310 |