missing translation for 'onlineSavingsMsg'
Learn More

Wijs solution, Volumetric, 0.1 M ICl, Honeywell Fluka™

Catalog No. 6001768
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001768 1 L
6001769 2.5 L
60046524 6 x 1 L
60046523 4 x 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6001768 Supplier Honeywell Chemicals Supplier No. 350711L
Only null left

Volumetric, 0.1 M ICl

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Chemical Identifiers

CAS 64-19-7 , 7790-99-0
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
PubChem CID 24640
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Quantity 1 L
Linear Formula ICl
MDL Number MFCD00036152
UN Number UN2920
Beilstein 3587194
Flash Point 40°C (104°F)
Synonym iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
Formula Weight 162.36g/mol
Chemical Name or Material Wijs solution
Hazard Statement P280-P305 + P351 + P338-P310