missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ UniStore™ Reagent Bottles, Wide Mouth, PP

Catalog No. S43631
Change view
Click to view available options
Capacity (Metric):
60 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S43631 60 mL
S43635 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S43631 Supplier United Scientific™ Supplier No. 33306

Wide Mouth Polypropylene Reagent Bottles are rigid, translucent and have fine contact clarity.

UniStore™ Wide Mouth, PP, reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant.

  • Excellent chemical resistance to most acids, bases, and alcohols.
  • Polypropylene caps are included.
  • Manufactured from USP Class VI resin that is FDA-21CFR compliant for food contact and free of heavy metals.
  • Leakproof closure with integrated seal ring.
  • Precision molded for consistent wall thickness with smooth interior and exterior surfaces.
  • Certificates of Analysis for leakproof test details are available upon request.
  • The 4000 mL bottle is leakproof at ambient temperature and pressure.

Specifications

Capacity 60 mL (2.03 oz.)
Capacity (Metric) 60 mL
Capacity (English) 2.03 oz.
Color Translucent
Diameter (Metric) Inner Neck 21 mm
Material Polypropylene
Quantity 12 Pk.
Sterility Non-sterile
Product Line UniStore
Closure Included Yes
Graduated No
Mouth Wide
Height (Metric) 10.2 cm
Product Type Reagent Bottle
Height (English) 4 in.
Closure Material Polypropylene
Closure Size 28 mm
Diameter (Metric) 12.7 cm
Through the Closure Dispensing No
Autoclavable Autoclavable
Show More Show Less