missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Wide Mouth Glass Reagent Bottle with Glass Stopper
Made from high quality borosilicate 3.3 glass. Extremely durable with a low temperature gradient and superior chemical resistance.
Supplier: Eisco™ CH0163C
Description
Warranty
1 year limited manufacturer warranty
Chemical Identifiers
| Clear | |
| Holding liquids | |
| Borosilicate Glass | |
| Clear | |
| Reusable | |
| Narrow | |
| Reagent Bottle | |
| Unlabeled | |
| Stopper |
| Wide neck | |
| Interchangeable, tight fitting, hollow glass stopper (socket size 24/29) | |
| Non-sterile | |
| Yes | |
| No | |
| Cylindrical | |
| Liquids | |
| Glass | |
| Autoclavable |
Specifications
| 250 mL | |
| Clear | |
| Holding liquids | |
| Borosilicate Glass | |
| Pack of 1 | |
| 317.51 g | |
| Clear | |
| Reusable | |
| Narrow | |
| 10.16 cm | |
| Liquids | |
| Unlabeled | |
| 34/35 mm | |
| Stopper |
| 8.5 oz. | |
| Wide neck | |
| Interchangeable, tight fitting, hollow glass stopper (socket size 24/29) | |
| 3.3 cm | |
| Non-sterile | |
| 3.3 cm | |
| Yes | |
| No | |
| Cylindrical | |
| Reagent Bottle | |
| 4 in. | |
| Glass | |
| 3.3 cm | |
| Autoclavable |