missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Wide Mouth Glass Reagent Bottle with Glass Stopper

Catalog No. S40894
Change view
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
2000 mL
Closure Size:
24/29 mm
29/32 mm
34/35 mm
45/40 mm
50/42 mm
55/44 mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
S40894 125 mL 29/32 mm
S40893 60 mL 24/29 mm
S40895 250 mL 34/35 mm
S40896 500 mL 45/40 mm
S40899 1000 mL 50/42 mm
S40901 2000 mL 55/44 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S40894 Supplier Eisco™ Supplier No. CH0163B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Made from high quality borosilicate 3.3 glass. Extremely durable with a low temperature gradient and superior chemical resistance.

  • Clear reagent bottle made from heavy duty borosilicate 3.3 glass
  • Wide mouth with interchangeable hollow borosilicate glass stopper
  • No graduations
  • Autoclavable
  • Warranty

    1 year limited manufacturer warranty

    Specifications

    Capacity (Metric) 125 mL
    Color Clear
    Description Wide neck
    For Use With (Application) Holding liquids
    Includes Interchangeable, tight fitting, hollow glass stopper (socket size 24/29)
    Material Borosilicate Glass
    Length (Metric) 3 cm
    Quantity 1
    Sterility Non-sterile
    Weight (Metric) 226.79 g
    Width (Metric) 3 cm
    Closure Color Clear
    Closure Included Yes
    Graduated No
    Mouth Narrow
    Height (Metric) 10.16 cm
    For Use With (Equipment) Liquids
    Height (English) 4 in.
    Chemical Label Unlabeled
    Closure Material Glass
    Closure Size 29/32 mm
    Diameter (Metric) 3 cm
    Closure Type Stopper
    Autoclavable Autoclavable
    Capacity (English) 4.2 oz.
    Shape Cylindrical
    Disposable Reusable
    Product Type Reagent Bottle
    Show More Show Less