missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Wide Mouth Glass Reagent Bottle with Glass Stopper

Catalog No. S40893
Change view
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
2000 mL
Closure Size:
24/29 mm
29/32 mm
34/35 mm
45/40 mm
50/42 mm
55/44 mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
S40893 60 mL 24/29 mm
S40894 125 mL 29/32 mm
S40895 250 mL 34/35 mm
S40896 500 mL 45/40 mm
S40899 1000 mL 50/42 mm
S40901 2000 mL 55/44 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S40893 Supplier Eisco™ Supplier No. CH0163A

Made from high quality borosilicate 3.3 glass. Extremely durable with a low temperature gradient and superior chemical resistance.

  • Clear reagent bottle made from heavy duty borosilicate 3.3 glass
  • Wide mouth with interchangeable hollow borosilicate glass stopper
  • No graduations
  • Autoclavable
  • Warranty

    1 year limited manufacturer warranty

    Specifications

    Capacity (Metric) 60 mL
    Capacity (English) 2 oz.
    Color Clear
    Description Wide neck
    For Use With (Application) Holding liquids
    Includes Interchangeable, tight fitting, hollow glass stopper (socket size 24/29)
    Material Borosilicate Glass
    Length (Metric) 2.5 cm
    Quantity 1
    Sterility Non-sterile
    Weight (Metric) 90.71 g
    Width (Metric) 2.5 cm
    Closure Color Clear
    Closure Included Yes
    Disposable Reusable
    Graduated No
    Mouth Narrow
    Shape Cylindrical
    Height (Metric) 10.16
    Product Type Reagent Bottle
    For Use With (Equipment) Liquids
    Height (English) 4 in.
    Chemical Label Unlabeled
    Closure Material Glass
    Closure Size 24/29 mm
    Diameter (Metric) 2.5 cm
    Closure Type Stopper
    Autoclavable Autoclavable
    Show More Show Less