Learn More
Thermo Scientific™ I-Chem™ Wide-Mouth Amber Glass Packer with Closure
GREENER_CHOICE
Collect soil, sludge, waste and water samples for a variety of organic and inorganic target analytes where light protection is required with these wide-mouth, amber glass packers.
$163.47 - $415.88
Specifications
Color Amber
Description Amber Glass Packer
Material Glass (Type 3)
Closure Color White
Liner Material PTFE
View More Specs
Products 18
Catalog Number Mfr. No. Capacity (Metric) Level Closure Size No. per Case Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Level Closure Size No. per Case Price Quantity & Availability  
05719289
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
141 0060
60 mL Unprocessed 33-400 mm 24
Case of 24 Each for $296.76
Only null left
05719288
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
241 0060
60 mL Processed 33-400 mm 24
Case of 24 Each for $350.26
Only null left
05719287
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
341 0060
60 mL Certified 33-400 mm 24
Case of 24 Each for $394.06
Only null left
05719291
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
341 0120
120 mL Certified 38-400 mm 12
Case of 12 Each for $219.65
Only null left
0571952
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
241 0120
120 mL Processed 38-400 mm 12
Case of 12 Each for $191.91
Only null left
05719292
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
141 0120
120 mL Unprocessed 38-400 mm 12
Case of 12 Each for $163.47
Only null left
0571966
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
341 0250
250 mL Certified 45-400 mm 12
Case of 12 Each for $238.85
Only null left
0571963
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
241 0250
250 mL Processed 45-400 mm 12
Case of 12 Each for $211.38
Only null left
05719294
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
141 0250
250 mL Unprocessed 45-400 mm 12
Case of 12 Each for $176.12
Only null left
0571969
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
141 0500
500 mL Unprocessed 53-400 mm 12
Case of 12 Each for $204.56
Only null left
0571974
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
241 0500
500 mL Processed 53-400 mm 12
Case of 12 Each for $227.18
Only null left
0571977
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
341 0500
500 mL Certified 53-400 mm 12
Case of 12 Each for $260.26
Only null left
0571979
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
141 0950
950 mL Unprocessed 53-400 mm 12
Case of 12 Each for $238.85
Only null left
0571980
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
241 0950
950 mL Processed 53-400 mm 12
Case of 12 Each for $279.74
Only null left
05719298
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
341 0950
950 mL Certified 53-400 mm 12
Case of 12 Each for $323.27
Only null left
05719467
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
341 1250
1250 mL Certified 70-400 mm 6
Case of 6 Each for $415.88
Only null left
05721106
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
141 2500
2500 mL Unprocessed 70-400 mm 4
Case of 4 Each for $243.15
Only null left
05721105
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
341 2500
2500 mL Certified 70-400 mm 4
Case of 4 Each for $278.56
Only null left
Description
Description
Collect soil, sludge, waste and water samples for a variety of organic and inorganic target analytes where light protection is required with Thermo Scientific™ I-Chem™ Wide-Mouth Amber Glass Packers. Packers are available Certified to meet EPA performance-based specifications for semi-volatile organics, pesticides, PCBs and metals anlyses, Processed the same way as the certified jars but without certification documentation, or Unprocessed and ready for your own cleaning procedure.
  • Amber Type III soda-lime glass jar; recommended for light-sensitive samples
  • PTFE-lined white polypropylene closed-top cap
  • Sizes range from 60 to 2500mL
  • Closures assembled on bottles
Certified jars come with the following features:
  • Certified to meet or exceed EPA performance-based standards for semi-volatile organics, pesticides, PCBs and metals analyses
  • Certificate of Analysis (CofA) included in the case and barcoded with lot number for traceability
  • Each jar individually barcoded with lot number and unique container number
  • Analysis labels included unattached for optional use
  • Carton sealed with security tape providing tamper evidence
  • Chipboard divider packaging to cushion and help prevent breakage; overpack required for shipping
Processed jars are processed in the same manner as certified jars and come with all the features of certified jars, but jars are not barcoded, and CofA is not included.
Unprocessed jars are ready for your own in-house cleaning and certification process. Boxed with chipboard dividers to help prevent breakage; overpack required for shipping.
Specifications
Specifications
Amber
Glass (Type 3)
PTFE
Amber Packer Bottle
Closed-top Cap
Amber Glass Packer
White
Wide
Polypropylene