missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ White/Tan PTFE/Silicone Septa
May be used in headspace solid-phase microextraction (SPME) (lyophilized aqueous extract)
Supplier: MilliporeSigma™ Supelco™ 27095U
Description
- A gas tight and inert nature septa, which is used for high temperature applications during pretreatment and sample oxidation.
Chemical Identifiers
| Tan, White | |
| PTFE/Silicone | |
| 100 |
Specifications
| 8 mm | |
| PTFE/Silicone | |
| Temperature limit 250°C | |
| 2 mL Standard Opening Vial |
| Tan, White | |
| 100 | |
| 0.06 in. |