missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ White/Tan PTFE/Silicone Septa

May be used in headspace solid-phase microextraction (SPME) (lyophilized aqueous extract)

Supplier:  MilliporeSigma™ Supelco™ 27177

 View more versions of this product

Catalog No. 111010683


Only null left

Description

Description

  • A gas tight and inert nature septa, which is used for high temperature applications during pretreatment and sample oxidation.
Specifications

Chemical Identifiers

Tan, White
PTFE/Silicone
100

Specifications

18 mm
PTFE/Silicone
Temperature limit 250°C
22 mL Vial
Tan, White
100
0.06 in. x 5 mil
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions