missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ White PTFE Cap Liner
Cap liners that can be used with Erlenmeyer flasks
Supplier: MilliporeSigma™ Supelco™ 27133
Description
- May be used for Erlenmeyer flasks for chemical oxygen demand (COD) determination.
- Oversized for better sealing ability
Chemical Identifiers
| Oversized for better sealing ability | |
| PTFE | |
| Temp. limit 225°C |
| White | |
| 100 | |
| 0.010 in. |
Specifications
| Oversized for better sealing ability | |
| White | |
| 100 | |
| 0.010 in. |
| 9 mm | |
| PTFE | |
| Temp. limit 225°C | |
| 2 mL Vial |