missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ White PTFE Cap Liner

Catalog No. 111011950
Change view
Click to view available options
Diameter (Metric) Inner:
9 mm
13 mm
15 mm
18 mm
21 mm
For Use With (Equipment):
15 mL Vial
2 mL Vial
22 mL Vial
4 mL Vial
7 mL Vial
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner For Use With (Equipment)
111011950 15 mm 7 mL Vial
111011951 18 mm 15 mL Vial
111012037 9 mm 2 mL Vial
111012039 13 mm 4 mL Vial
111012065 21 mm 22 mL Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 111011950 Supplier MilliporeSigma™ Supelco™ Supplier No. 27157

Cap liners that can be used with Erlenmeyer flasks

  • May be used for Erlenmeyer flasks for chemical oxygen demand (COD) determination.
  • Oversized for better sealing ability

Specifications

Description Oversized for better sealing ability
Diameter (Metric) Inner 15 mm
Color White
Material PTFE
No. per Pack 100
Temperature Range (English) Temp. limit 225°C
Thickness (English) 0.010 in.
For Use With (Equipment) 7 mL Vial