missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ White Biohazard Suit with Elastic Wrists and Ankles
These disposable coveralls are designed with fabric and seams that have been rigorously tested for protection against blood and blood-borne pathogens.
Supplier: International Enviroguard™ W2502L
Description
- Both the fabric and seams pass ASTM F1670 and F1671 tests for protection against blood, bloodborne pathogens, and bodily fluids
- Durable fabric is built to withstand tears and abrasions in tough working environments
- Zipper front with a sealable storm flap helps keep contaminants from entering or seeping through exposed zipper seams
- Red fabric pull tab on the zipper makes it quick and easy to find so you can suit up or down in no time
- Taped seams help keep contaminants out for added protection, while elastic wrists and ankles keep coveralls secure and in place
- Convenient thumb and finger loops help keep sleeves from riding up while you work
- Double-sided tape around wrists helps keeps gloves in place during donning
- Storm flap over zipper helps prevent contaminants from entering through otherwise exposed zipper seams
- Roomy one-piece raglan sleeves resist tears around shoulders
- A gusseted crotch gives the user more freedom to squat, bend, and crouch without ripping fabric
- Possible applications include emergency response, healthcare, category 3 black water, crime scene remediation, and animal research
Specifications
| A50 Breathable Particle and Splash Protection Coveralls | |
| Unisex | |
| White | |
| Elastic at Back | |
| Elastic, Finger Loops | |
| Standard | |
| Serged, Taped | |
| Bloodborne Pathogens, Particle, Liquid, Tears | |
| None |
| Large | |
| Multi-layer Barrier Laminate Fabric | |
| Single-use | |
| Elastic | |
| Collared | |
| 2-way Zipper with Sealed Storm Flap | |
| Non-sterile | |
| Yes |