missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ White Biohazard Suit with Elastic Wrists and Ankles
These disposable coveralls are designed with fabric and seams that have been rigorously tested for protection against blood and blood-borne pathogens.
$629.35 - $765.05
Specifications
| Product Type | A50 Breathable Particle and Splash Protection Coveralls |
|---|---|
| Gender | Unisex |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | White |
| Disposable | Single-use |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
17-183-948
|
International Enviroguard™
W2502S |
Small |
Case of 25 Each for $629.35
|
|
|||||
|
17-183-949
|
International Enviroguard™
W2502M |
Medium |
Case of 25 Each for $629.35
|
|
|||||
|
17-183-950
|
International Enviroguard™
W2502L |
Large |
Case of 25 Each for $629.35
|
|
|||||
|
17-183-947
|
International Enviroguard™
W2502XL |
X-Large |
Case of 25 Each for $629.35
|
|
|||||
|
17-183-953
|
International Enviroguard™
W25022XL |
2X-Large |
Case of 25 Each for $662.84
|
|
|||||
|
17-183-952
|
International Enviroguard™
W25023XL |
3X-Large |
Case of 25 Each for $704.35
|
|
|||||
|
17-183-951
|
International Enviroguard™
W25024XL |
4X-Large |
Case of 25 Each for $765.05
|
|
|||||
Description
- Both the fabric and seams pass ASTM F1670 and F1671 tests for protection against blood, bloodborne pathogens, and bodily fluids
- Durable fabric is built to withstand tears and abrasions in tough working environments
- Zipper front with a sealable storm flap helps keep contaminants from entering or seeping through exposed zipper seams
- Red fabric pull tab on the zipper makes it quick and easy to find so you can suit up or down in no time
- Taped seams help keep contaminants out for added protection, while elastic wrists and ankles keep coveralls secure and in place
- Convenient thumb and finger loops help keep sleeves from riding up while you work
- Double-sided tape around wrists helps keeps gloves in place during donning
- Storm flap over zipper helps prevent contaminants from entering through otherwise exposed zipper seams
- Roomy one-piece raglan sleeves resist tears around shoulders
- A gusseted crotch gives the user more freedom to squat, bend, and crouch without ripping fabric
- Possible applications include emergency response, healthcare, category 3 black water, crime scene remediation, and animal research
Specifications
| A50 Breathable Particle and Splash Protection Coveralls | |
| Multi-layer Barrier Laminate Fabric | |
| Single-use | |
| Elastic | |
| Standard | |
| Serged, Taped | |
| Bloodborne Pathogens, Liquid, Particle, Tears | |
| None |
| Unisex | |
| White | |
| Elastic at Back | |
| Collared | |
| 2-way Zipper with Sealed Storm Flap | |
| Non-sterile | |
| Yes |