Learn More
DWK Life Sciences Wheaton™ White Polypropylene Caps with Polyvinyl Liners
Good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
$77.21 - $146.07
Specifications
Color White
Material Polypropylene
View More Specs
Products 9
Catalog Number Mfr. No. Neck Finish No. per Case Price Quantity  
Catalog Number Mfr. No. Neck Finish No. per Case Price Quantity  
02-984-54
View Documents
DWK Life Sciences
239220
70-400 48 Cs.
Case of 48 Each for $146.07
Only null left
02-984-42
View Documents
DWK Life Sciences
239216
48-400 72 Cs.
Case of 72 Each for $104.86
Only null left
02-984-21
View Documents
DWK Life Sciences
239209
24-400 144 Cs.
Case of 144 Each for $100.79
Only null left
02-984-33
View Documents
DWK Life Sciences
239213
38-400 72 Cs.
Case of 72 Each for $77.21
Only null left
02-984-48
View Documents
DWK Life Sciences
239218
58-400 72 Cs.
Case of 72 Each for $128.29
Only null left
02-984-17
View Documents
DWK Life Sciences
239207
22-400 144 Cs.
Case of 144 Each for $115.21
Only null left
02-984-60
View Documents
DWK Life Sciences
239222
89-400 48 Cs.
Case of 48 Each for $135.14
Only null left
02-984-25
View Documents
DWK Life Sciences
239211
28-400 144 Cs.
Case of 144 Each for $106.25
Only null left
02-984-45
View Documents
DWK Life Sciences
239217
53-400 72 Cs.
Case of 72 Each for $111.71
Only null left
Description
Description
  • White polypropylene screw cap provides clear surface for writing on cap
  • Poly-vinyl liner provides an excellent moisture barrier
Specifications
Specifications
White
Polypropylene
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'