missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Caps with Polyvinyl Liners

Catalog No. 0298454
Change view
Click to view available options
Neck Finish:
22-400
24-400
28-400
38-400
48-400
53-400
58-400
70-400
89-400
No. per Case:
144 Cs.
48 Cs.
72 Cs.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case
0298454 70-400 48 Cs.
0298442 48-400 72 Cs.
0298421 24-400 144 Cs.
0298433 38-400 72 Cs.
0298448 58-400 72 Cs.
0298417 22-400 144 Cs.
0298460 89-400 48 Cs.
0298425 28-400 144 Cs.
0298445 53-400 72 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 0298454 Supplier DWK Life Sciences Supplier No. 239220

Good for mild acids, alkalis, solvents, alcohols, oils and aqueous products

  • White polypropylene screw cap provides clear surface for writing on cap
  • Poly-vinyl liner provides an excellent moisture barrier

Specifications

Color White
Material Polypropylene
No. per Case 48 Cs.
Neck Finish 70-400
Certifications/Compliance ISO 9001
Product Type Polypropylene Cap
For Use With (Equipment) Round bottle
Closure Liner Polyvinyl