Learn More
DWK Life Sciences Wheaton™ Viton™ Tubing Assemblies
Resistant to oils, chemicals, weather-related aging and ozone
Supplier: DWK Life Sciences 374316
Description
For Unispense peristaltic pumps
- Material retains its good mechanical and chemical resistance properties at extremely high temperatures better than any other known elastomer
- Tubing assembly consists of 5 ft. (1.5 m) section of silicone or FKM tubing with a wall thickness of 2.3 mm and borosilicate glass sinker on one end and a polypropylene delivery nozzle on the other end
Chemical Identifiers
| Viton | |
| 150 cm | |
| 2.3 mm | |
| Tubing Assembly |
| 4.92 ft. | |
| 0.09 in. | |
| Borosilicate Glass sinker on one end and Polypropylene delivery nozzle on the other end |
Specifications
| 11/100 in. | |
| Viton | |
| 150 cm | |
| 2.3 mm | |
| Unispense™ peristaltic pump | |
| Tubing Assembly |
| 3 mm | |
| 4.92 ft. | |
| 0.09 in. | |
| Unispense™ peristaltic pump | |
| Borosilicate Glass sinker on one end and Polypropylene delivery nozzle on the other end |