Learn More
DWK Life Sciences Wheaton™ V Vial™ with Solid-Top Screw Cap, Clear
Conical interior bottoms are designed to provide total downward drainage and maximum retrieval of contents by syringe
Supplier:  DWK Life Sciences W986276NG
 View more versions of this product
Catalog No. 03410029
Only null left
Includes: 14B rubber-lined black phenolic screw caps faced with PTFE
Description
Description
Clear vials are manufactured from Wheaton-33™ low extractable borosilicate glass
Specifications
Chemical Identifiers
Clear
Ideal for small scale reactions, centrifugation, storage, packaging and shipping
Screw
Screw Cap
Borosilicate Glass (Low Extractable)
Conical V
Phenolic
ASTM E438, USP Type I, Type I, Class A
Specifications
PTFE/14B Rubber
17 mm
2.0 mL
Yes
Screw
Phenolic
15-415
61 mm
Clear
Borosilicate Glass (Low Extractable)
Ideal for small scale reactions, centrifugation, storage, packaging and shipping
Conical V
0.5 dr.
Screw Cap
ASTM E438, USP Type I, Type I, Class A
Product Suggestions
Product Suggestions