Learn More
DWK LIFE SCIENCES™ Socorex™ Twister™ Universal Pipette Work Station
Catalog No. 1370985
Change view
Click to view available options
Color:
Blue
Gray
Green
Orange
Red
White
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
1370985 Yellow
1370980 Green
1370982 Gray
1370981 Orange
1370979 Blue
1370984 White
1370983 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 1370985 Supplier DWK LIFE SCIENCES™ Supplier No. W870164
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Twister™ Universal Pipette Work Stations from DWK LIFE SCIENCES™ feature 360° rotation for easy access to each pipette. Holds up to six pipettes of most brands. Easy to disassemble for cleaning and maintenance. Available in multiple translucent colours with exchangeable discs.
  • Twister™ Universal Pipette Work Station designed for organized, efficient pipette storage on the benchtop
  • 360° rotation allows easy access to each pipette on the stand
  • Can hold up to six pipettes of most brands
  • Can be fully disassembled and cleaned for easy maintenance
  • Available in multiple translucent colors for color-coding and lab organization
Specifications
Autoclavable Not Autoclavable
Color Yellow
Holds Pipets
Material Polyamide
Positions 6
Stackable No
Sterility Non-sterile
Product Type Pipet Stand
Shelves None
Height (Metric) 220 mm
Width (Metric) 144 mm
Show More Show Less