missing translation for 'onlineSavingsMsg'
Learn More

Socorex™ Acura™ Single-Channel Variable Volume Micropipetters from DWK LIFE SCIENCES™

Catalog No. 13709402
Change view
Click to view available options
Volume (Metric):
0.1 to 2 μL
0.5 to 10 μL
1 to 10 μL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
5 to 50 μL
Model:
825
826 XS
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Model
13709402 0.5 to 10 μL 826 XS
1370933 0.5 to 10 μL 825
1370932 0.1 to 2 μL 825
1370934 1 to 10 μL 825
1370935 2 to 20 μL 825
1370936 5 to 50 μL 825
1370937 10 to 100 μL 825
1370938 20 to 200 μL 825
1370939 100 to 1000 μL 825
13709401 0.1 to 2 μL 826 XS
13709403 1 to 10 μL 826 XS
13709404 2 to 20 μL 826 XS
13709406 10 to 100 μL 826 XS
13709405 5 to 50 μL 826 XS
13709407 20 to 200 μL 826 XS
13709408 100 to 1000 μL 826 XS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 13709402 Supplier Socorex™ Supplier No. W810302XS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Feature swift-set calibration requiring no additional tools, reducing downtime, fully autoclavable to 121°C, ergonomic design, and JustTip™ system ensure precise, comfortable pipetting

  • Fully assembled unit is autoclavable to 121°C (250°F), reducing the time needed to bring pipetter back into service after contamination
  • Tip ejector is positioned for easy use by either hand, even when wearing gloves
  • JustTip™ system allows for easy adjustment of tip ejector so that it can be used with most tips
  • Volume display can be easily read without changing hand position
  • Excellent ergonomic shape and size ratio; very light weight
  • New springs and tightness seal for extra-smooth activation
  • Increased drivability with short, narrow shaft
  • Easier to use when working in a fume hood
  • Conical shaft end for easy reach in smaller microtubes

Warranty

Three years

Specifications

Accuracy ±1%, <±1.8%, <±2.5%
Certifications/Compliance CE, IVD 98/79 EEC
Compatible Tips Ultra 10μL tip
Autoclavable Autoclavable
Color Blue
Disposable Reusable
For Use With Ultra Pipette Tip (10 μL)
Grip Type Ergonomic
Material Plastic
Model 826 XS
Number of Channels 1
Product Line Acura
Programmability No
Sterility Non-sterile
Type Variable Volume Micropipette
Volume (Metric) 0.5 to 10 μL
Finger Style Ergonomic
For Use With (Equipment) Ultra 10μL tip
Increments 0.01μL
Secure Tip Attachment JusTip™ adjustable tip ejection
Warranty 3 Years
Show More Show Less