Learn More
Socorex Acura™ Single-Channel Variable Volume Micropipetters from DWK Life Sciences™
Swift-set calibration requires no additional tools, reduces downtime
Supplier:  Socorex W810306
 View more versions of this product
Catalog No. 13-709-35
Only null left
Explore available promotions
Description
Description
Fully assembled unit is autoclavable to 121°C (250°F), reducing the time needed to bring pipetter back into service after contamination.
  • Tip ejector is positioned for easy use by either hand, even when wearing gloves
  • JustTip™ system allows for easy adjustment of tip ejector so that it can be used with most tips
  • Volume display can be easily read without changing hand position
  • Excellent ergonomic shape and size ratio; very light weight
  • New springs and tightness seal for extra-smooth activation
  • Increased drivability with short, narrow shaft
  • Easier to use when working in a fume hood
  • Conical shaft end for easy reach in smaller microtubes
Warranty
Three years
Specifications
Specifications
±1%, <±1.8%, <±2.5%
Micro 200μL tip
Micro Tip 200 μL
1
Variable Volume Micropipette
Ergonomic
0.02μL
3 Years
CE, IVD 98/79 EEC
Autoclavable
825
Acura
2 to 20 μL
Micro 200μL tip
JusTip™ adjustable tip ejection
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories