Learn More
Socorex Acura™ Manual Pipets 826 XS TwiXS pack from DWK Life Sciences™
Catalog No. 13710911
Change view
Click to view available options
Volume (Metric):
0.1 to 2, 2 to 20 μL
0.5 to 10, 10 to 100 μL
10 to 100, 100 to 1000 μL
2 to 20, 20 to 200 μL
20 to 200, 100 to 1000 μL
5 to 50, 20 to 200 μL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
13710911 20 to 200, 100 to 1000 μL
13709409 0.1 to 2, 2 to 20 μL
13709410 0.5 to 10, 10 to 100 μL
13709411 5 to 50, 20 to 200 μL
13709412 10 to 100, 100 to 1000 μL
13710910 2 to 20, 20 to 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 13710911 Supplier Socorex Supplier No. W810315TXS
Includes
Shelf pipette holder, pipette tips samples, silicone grease, Q.C. certificates and operating instructions
Two adjustable volume pipettes available in six different packs
  • Two adjustable volume pipettes
  • Free shelf pipette holder included
  • Volume combinations up to 1000μL
  • Six different packs available
Specifications
Autoclavable Autoclavable
Model 826 XS
Product Line Acura
Type Pipette
Volume (Metric) 20 to 200, 100 to 1000 μL
Finger Style Ergonomic
Includes Shelf pipette holder, pipette tip samples, Silicone Grease, Q.C. certificates and operating instructions
UV Monitoring Yes
Warranty 3 Years