Learn More
Socorex Acura™ Manual Pipets 826 XS TwiXS pack from DWK Life Sciences™
Two adjustable volume pipettes available in six different packs
Supplier:  Socorex W810311TXS
 View more versions of this product
Catalog No. 13709412
Only null left
Includes: Shelf pipette holder, pipette tips samples, silicone grease, Q.C. certificates and operating instructions
Description
Description
  • Two adjustable volume pipettes
  • Free shelf pipette holder included
  • Volume combinations up to 1000μL
  • Six different packs available
Specifications
Chemical Identifiers
CE, IVD 98/79 EEC
826 XS
Ergonomic
Yes
Autoclavable
Acura
Shelf pipette holder, pipette tip samples, Silicone Grease, Q.C. certificates and operating instructions
3 Years
Specifications
CE, IVD 98/79 EEC
826 XS
Pipette
Ergonomic
Yes
Autoclavable
Acura
10 to 100, 100 to 1000 μL
Shelf pipette holder, pipette tip samples, Silicone Grease, Q.C. certificates and operating instructions
3 Years
Product Suggestions
Product Suggestions