Learn More
DWK Life Sciences Wheaton™ Serum Bottles and Vials
These bottles have exceptional wall strength and superior heat and chemical tolerances
Supplier:  DWK Life Sciences 223738
 View more versions of this product
Catalog No. 06406C
Only null left
Description
Description
These bottles have exceptional wall strength and superior heat and chemical tolerances
Ideal for long- and short-term sample storage and vaccine/injectable drug containers.
  • Clear borosilicate glass conforms to USP Type 1 requirements
  • Amber borosilicate glass conforms to USP Type 1 requirements
  • Shrink wrapped partitioned modules reduce breakage
Compliance
Meet requirements for Class A of the United States Pharmacopeia for Type I glass.
Specifications
Chemical Identifiers
Borosilicate Glass
Specifications
0.17 oz., 5mL
0.17 oz.
0.9 in.
47 mm
23 mm
5 mL
Clear
Borosilicate Glass
1.85 in.
Product Suggestions
Product Suggestions