Learn More
DWK Life Sciences Wheaton™ Flat Bottom Flasks For Soxhlet Extraction Apparatus
Catalog No. 02912579
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02912579 500 mL
02912577 125 mL
02912578 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02912579 Supplier DWK Life Sciences Supplier No. 428164
Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
  • Flat bottom flask 24/42
Specifications
Capacity (Metric) 500 mL
Material Borosilicate Glass
For Use With (Application) For continuous extraction of analytes from a solid into an organic solvent
Standard Taper Joint 24/40
Bottom Shape Flat