Learn More
DWK Life Sciences Wheaton™ Safety-Coated Media Bottles
Create a safe work environment with Wheaton Safety Coated Bottles. If a bottle is accidentally broken, the PVC based plastisol coating on the outside of bottle retains the contents long enough for proper disposal.
$1,045.32
Specifications
Coating Plastisol
Color Clear
Material Borosilicate Glass
Graduated Yes
Product Type Media Bottle
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
06451274
View Documents Promotion Details
DWK Life Sciences
219455
125 mL
Case of 48 Each for $1,045.32
Only null left
Description
Description
  • USP Type I clear borosilicate glass
  • Plastisol coating for safety
  • Provided with graduations and writing patch
  • 33-430 screw cap can be purchased separately
  • Autoclavable
Specifications
Specifications
Plastisol
Borosilicate Glass
Media Bottle
Screw Cap
Clear
Yes
33-430 mm