Learn More
DWK Life Sciences Wheaton™ Redi-Cap™ Phenolic Screw Caps for Redi-Pak™ Containers
Unique cone liner design offers superior torque retention and excellent sealing characteristics
Supplier:  DWK Life Sciences 239255
 View more versions of this product
Catalog No. 02-984-19
Only null left
Explore available promotions
Description
Description
  • Poly-Seal liner is manufactured from low density polyethylene
  • Phenolic
  • Screw cap
  • Color: Black
Specifications
Specifications
Black
144 Cs.
ISO 9001
Polyseal
Phenolic Resin
22-400
Redi-pak™ container
Product Suggestions
Product Suggestions