Learn More
DWK Life Sciences Wheaton™ PP Made Delivery Nozzles
Polypropylene Nozzle and Glass Sinker - nozzle connects to the pump tubing for dispensing of liquids
$170.64 - $192.61
Specifications
Autoclavable Autoclavable
Material Polypropylene
For Use With Wheaton peristaltic pumps, dispensing pumps and tubing
Product Type Delivery Nozzle
View More Specs
Products 3
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
02913316
View Documents Promotion Details
DWK Life Sciences
374328
8 mm
Case of 6 Each for $170.64
Only null left
02913314
View Documents Promotion Details
DWK Life Sciences
374324
2 and 3 mm
Case of 6 Each for $192.61
Only null left
02913315
View Documents Promotion Details
DWK Life Sciences
374326
6 mm
Case of 6 Each for $170.64
Only null left
Description
Description
  • Glass sinker is used to secure tubing when drawing from a container
Specifications
Specifications
Autoclavable
Polypropylene
Wheaton peristaltic pumps, dispensing pumps and tubing
Delivery Nozzle