Learn More
DWK Life Sciences Wheaton™ PP Made Delivery Nozzles
Polypropylene Nozzle and Glass Sinker - nozzle connects to the pump tubing for dispensing of liquids
Supplier:  DWK Life Sciences 374326
 View more versions of this product
Catalog No. 02913315
Only null left
Description
Description
  • Glass sinker is used to secure tubing when drawing from a container
Specifications
Chemical Identifiers
Autoclavable
Polypropylene
Wheaton peristaltic pumps, dispensing pumps and tubing
Delivery Nozzle
Specifications
6 mm
Polypropylene
Delivery Nozzle
Autoclavable
Wheaton peristaltic pumps, dispensing pumps and tubing
Product Suggestions
Product Suggestions