Learn More
DWK Life Sciences Wheaton™ Potter-Elvehjem Tissue Grinders With Radial Serrations
Serrated PTFE pestle disperses homogenate into the mortar cylinder more efficiently
Supplier:  DWK Life Sciences 357994
 View more versions of this product
Catalog No. 02-911-528
Only null left
Explore available promotions
Description
Description
Grinders are precisely constructed with completely interchangeable parts, allowing for easier replacement of parts.
  • Mortar made from low extractable borosilicate glass
  • Radial serrated PTFE pestle
  • Interchangeable parts
  • Autoclavable
Order Info
Grinding chamber clearance is 0.004 in. to 0.006 in. (0.1 to 0.15mm). Depth of the serrations is 0.020 in. to .025 in. . Diameter of the stainless steel shaft is 0.25 in. (6.3mm).
Specifications
Specifications
1.18 in.
1.85 oz.
Borosilicate Glass, PTFE
10.4 in.
5.1 in.
1.85 oz., 55mL
30mm
55 mL
Tissue Grinder
266mm
130mm
Product Suggestions
Product Suggestions