missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ PET Diagnostic Bottles

Catalog No. 12141614
Change view
Click to view available options
Color:
Amber
Clear
Capacity (Metric):
5 mL
20 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Capacity (Metric)
12141614 Amber 20 mL
12141613 Clear 5 mL
12141607 - 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12141614 Supplier DWK Life Sciences Supplier No. W220005

These bottles provide a lightweight, transparent solution for storing reagents and buffer solutions. Available in clear or amber, they offer easy content visibility and light protection, with a pre‐attached screw cap and liner for secure, reliable closure.

  • Polyethylene Terephthalate, transparent
  • Ideal for reagents and buffer solutions
  • Clear and amber versions available
  • Pre-attached white polypropylene screw cap with foamed polyethylene liner

Specifications

Capacity (Metric) 20 mL
Color Amber
For Use With (Application) Diagnostic Reagent and Buffer Storage
Includes Screw Caps
Height (Metric) With Closure 65.5 mm
Material PET
Quantity 100
Sterility Non-sterile
Thread Size (Metric) 20-410
Closure Included Yes
Graduated No
Mouth Narrow, Threaded
Fit for Purpose Certified Quality
Height (Metric) 62.5 mm
Chemical Label Unlabeled
Closure Liner F217 Foamed PE
Closure Material Polypropylene
Closure Size 20 mm
Closure Type Screw Cap
Diameter (Metric) Outer Tubing 29.7 mm
Autoclavable Not Autoclavable
Capacity (English) 0.68 fl oz.
Product Type Reagent Bottle
Show More Show Less