Learn More
DWK Life Sciences Wheaton™ Leak-Resistant Narrow-Mouth Natural PP Bottles
With double-seal closure with cap attached
$218.06 - $605.64
Specifications
Color Natural
Includes Double seal closure, no-drip pour lip
Material Polypropylene
Packaging Convenience
Certifications/Compliance ISO 9001
View More Specs
Products 9
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Qty  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Qty  
03-083-26
missing translation for 'viewDocuments'
DWK Life Sciences
209161
4 mL 13-425 mm
Case of 72 Each for $218.06
Only null left
03-083-27
missing translation for 'viewDocuments'
DWK Life Sciences
209163
8 mL 20-410 mm
Case of 72 Each for $244.09
Only null left
03-083-28
missing translation for 'viewDocuments'
DWK Life Sciences
209164
15 mL 20-410 mm
Case of 72 Each for $293.36
Only null left
03-083-29
missing translation for 'viewDocuments'
DWK Life Sciences
209165
30 mL 20-410 mm
Case of 72 Each for $290.09
Only null left
03-083-30
missing translation for 'viewDocuments'
DWK Life Sciences
209166
60 mL 20-410 mm
Case of 72 Each for $288.79
Only null left
03-083-31
missing translation for 'viewDocuments'
DWK Life Sciences
209167
125 mL 24-410 mm
Case of 72 Each for $439.03
Only null left
03-083-32
missing translation for 'viewDocuments'
DWK Life Sciences
209168
250 mL 24-410 mm
Case of 72 Each for $605.64
Only null left
03-083-33
missing translation for 'viewDocuments'
DWK Life Sciences
209169
500 mL 28-410 mm
Case of 48 Each for $599.91
Only null left
03-084-00
missing translation for 'viewDocuments'
DWK Life Sciences
209170
1000 mL 38-430 mm
Case of 24 Each for $446.15
Only null left
Description
Description
Ideal for short- and long-term storage and packaging.
  • No drip pour lip for clean pouring
  • Polypropylene cap attached remains tight over an extended time
  • Convenience packed
Specifications
Specifications
Natural
Polypropylene
ISO 9001
Round
Screw Cap
Double seal closure, no-drip pour lip
Convenience
Narrow
Polypropylene