missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ HPLC Mobile-Phase Reservoirs
Suitable for use as mobile phase and filtration reservoirs, as well as general sample and buffer storage
$654.25 - $1383.93
Specifications
| Certifications/Compliance | USP Type I and ASTM E 438 Type I, Class A |
|---|---|
| Material | Glass |
| Type | HPLC Mobile-phase Reservoir |
| For Use With (Application) | Ideal for mobile phase or filtration reservoir |
| Bottom Shape | Flat |
| Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) Outer | Height (Metric) | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) Outer | Height (Metric) | Price | Quantity | |||
|
13-703-993
|
DWK Life Sciences
264710 |
2000 mL | 138 mm | 246 mm |
Case for $654.25
|
|
|||
|
13-703-994
|
DWK Life Sciences
264711 |
5000 mL | 186 mm | 318 mm |
Each for $1,383.93
|
|
|||
Description
- Manufactured from Wheaton 33™ low extractable borosilicate glass, which meets ASTM Type I, Class A and USP Type I standards
- Plastic safety coating with an integral UV-blocking agent helps retain glass and liquid in case of breakage and block virtually all UV light up to 385nm
- Screw thread: GL 45
- Autoclavable up to 121°C at 15-20 psig for 15-20 minutes
- Flat bottom
Specifications
| USP Type I and ASTM E 438 Type I, Class A | |
| HPLC Mobile-phase Reservoir | |
| Flat |
| Glass | |
| Ideal for mobile phase or filtration reservoir |