missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ Glass Staining Dishes for 50 Slides

Catalog No. 22031300
Change view
Click to view available options
Includes:
Cover
Dish
Dish, Cover
Dish, Cover, Rack
Rack
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22031300 Cover
08813A Dish, Cover, Rack
08813C Dish, Cover
22029567 Dish
22029568 Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22031300 Supplier DWK Life Sciences Supplier No. 900402

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

WHEATON™ Glass Staining Dishes for 50 Slides are made from soda-lime glass and hold up to 50 standard slides. The stainless steel rack is nontarnishable, resists staining solutions, and features a hinged handle for easy slide insertion and removal.

  • Dish and cover are made from soda-lime glass
  • Holds up to 50 standard slides ( 3 x 1, 3 x 1.5, 3 x 2 in.)
  • Stainless steel rack is nontarnishable and withstands most staining solutions
  • Hinged handle allows easy insertion and removal of slides
  • Open-bottom rack design facilitates rapid immersion and drainage, reducing carryover
  • Slides can be placed singly, alternately straight across and diagonally, or back-to-back
  • Dimensions: 185 L x 88 W x 78 mm D
  • Available as complete unit or as individual components (dish, cover, rack)

Specifications

Closure Included Yes
Autoclavable Not Autoclavable
Bottom Shape Flat
Color Clear
Material Soda Lime Glass
Type Cover
For Use With (Equipment) Glass staining dish for 50 slides
Includes Cover