missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Glass Serum Bottles

Catalog No. p-45470
Change view
Click to view available options
Capacity (Metric):
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
01000800 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01000800 Supplier DWK Life Sciences Supplier No. 223749
Only null left

WHEATON™ Glass Serum Bottles are ideal for long term sample storage and drug containers.

  • WHEATON™ Glass Serum Bottles are manufactured from Type I Borosilicate molded glass that conforms to USP Type I requirements.
  • Raised graduations indicate approximate capacities in milliliters.
  • WHEATON™ Glass Serum Bottles accept standard stoppers and seals.
  • The bottles are non-graduated.
    • Specifications

      Capacity (Metric) 250 mL
      Capacity (English) 8.45 oz.
      Color Clear
      Diameter (English) 2.36 in.
      Diameter (Metric) Outer Neck 30 mm
      Diameter (Metric) Inner Neck 15 mm
      Material Borosilicate Glass
      No. per Case 35 Cs.
      Height (Metric) 158 mm
      Product Type Serum Bottle
      Height (English) 6.22 in.
      Diameter (Metric) 60 mm
      Show More Show Less